Hydrochloric acid 0.1 mol/l (0.1 N) in 2-propanol acc. to DIN 51558 part 1 , AVS TITRINORM
Catalog # VWRC86170.290
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pk:1 L
- Seq:5037
- Environmentally Preferable:
Specifications
Specification Test Results
- Titer (20°C) (result of analysis at point of filling):0.9980 – 1.0020
Specification Test Results
About this item
Synonyms: Hydrochloric acid solution, Muriatic acid
:
ADR:8,I
:36.46 g/mol
UN:2920
:GHS05
:GHS02
:GHS07
Storage Temperature:Ambient
CAS Number:7647-01-0
Formula:HCl
MDL Number:MFCD00011324