Order Entry
China
ContactUsLinkComponent
Supelco® Organic Reference Standard, Triglyceride Mix
Supelco® Organic Reference Standard, Triglyceride Mix
  SUPL17810-1AMP-S
 :  SUPELCO
 :  
Supelco® Organic Reference Standard, Triglyceride Mix
  SUPL17810-1AMP-S
 :  SUPELCO
 :  

 

  • Pk:
    1 Ampoul
  • Description:
    Triglyceride Mix
  • Storage conditions:
    –10...–25 °C
  • Shipping conditions:
    DICE
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    Lipid standards: triglyceride mixtures
  • Storage temperature:
    −20 °C
  • Flash point:
    148,9 °C
  • Pk Info:
    Ampoule of 100 mg (Neat)

 

 

 : 100 mg ampule contains approximately equal amounts by weight of: Triacetin (C2:0) Tributyrin (C4:0) Tricaproin (C6:0) Tricaprylin (C8:0) Tricaprin (C10:0)