Organic reference standards, D-alpha-Tocopherol succinate, analytical standard, Supelco®
目录编号 SUPL47782
Supplier: SUPELCO
CAS Number:
Specifications
- Pack type:Ampule
- Pk:1
- Standards:Analytical standard
- CAS No.:4345-03-3
- Description:D-α-Tocopherol succinate
- Formula:C₃₃H₅₄O₅
- Storage temperature:–20 °C
- Description:D-α-Tocopherol succinate
- Pk Info:100 mg
- Form:Neat