Organic reference standard, Butylated hydroxyanisole, analytical standard, Supelco®
Catalog # SUPL78943-500MG
Supplier: SUPELCO
CAS Number:
Specifications
- Pk:500 mg
- Standards:Analytical standard
- CAS No.:25013-16-5
- Description:Butylated hydroxyanisole, ≥99,0% (sum of isomers, GC)
- Melting point:58...60 °C (lit.)
- Formula:C₁₁H₁₆O₂
- Description:Butylated hydroxyanisole
- Form:Neat
Specifications
About this item
Butylated hydroxyanisole is a lipophilic compound, which is is widely used as an antioxidant in food industry.