Order Entry
China
ContactUsLinkComponent
Organic reference standard, (+/-)-1,3-Butanediol, analytical standard, Supelco®
Organic reference standard, (+/-)-1,3-Butanediol, analytical standard, Supelco®
Catalog # SUPL09068-1ML
Supplier:  SUPELCO
CAS Number:  
undefined
Organic reference standard, (+/-)-1,3-Butanediol, analytical standard, Supelco®
Catalog # SUPL09068-1ML
Supplier:  SUPELCO
CAS Number:  

Specifications

  • Pk:
    1 mL
  • Standards:
    Analytical standard
  • CAS No.:
    107-88-0
  • Description:
    (+/-)-1,3-Butanediol, ≥99,0% (GC)
  • Seq:
    01
  • Formula:
    C₄H₁₀O₂
  • Description:
    (+/-)-1,3-Butanediol
  • Boiling point:
    203...204 °C (lit.)

Specifications

About this item

(±)-1,3-Butanediol is an organic alcohol with antimicrobial activity against a wide variety of pathogens.

  • Used as a reference standard in the determination of (±)-1,3-butanediol in biological samples using wide-bore column gas chromatography coupled to flame ionization detector.