Cut resistant gloves, ChainExtra, with short cuff
Supplier: HONEYWELL SAFETY PRODUCTS
Certificates
About this item
These fully reversible chainmail gloves are equipped with heavy duty adjustable straps and press stud fastening. They combines safety, quality and comfort, protecting against cuts, a common hazard in the meat and processing industries.
- Detachable and interchangeable plastic straps with antimicrobial agent
- Damaged straps can be changed over quickly and easily
- Firm fastening for a secure fit every time; there is no side slit which may allow a knife to enter at wrist level
- Fitted with an identification tag which can be personalised by the wearer
- Each glove is identified by a unique number for traceability and record keeping
- Exceptional quality of chain mesh welds, reducing the number of repairs, and increasing the life time of the glove
- Protection for hand and part of forearm
Typical applications include: Metal works, meat processing lines, kill floors, meat deboning, poultry works, fish processing.
Length: 308 to 370 mm (depending on size)
Weight: 254 to 360 g (depending on size)
Chainmail thickness: 0,05 mm
External diameter of ring: 4 mm
Certifications: EN 14328; Conforms with EC directive 89/686/CEE
Packaging: Every glove is packaged in a bag or a case on which is pasted a tag indicating the model, together with a multilingualinstruction for use.
Caution: The colour given in the ordering table states the colour of the colour-coded plastic straps.
Specifications
- Catalog No:
- DALL2542414R0302
- DALL2542413R0302
- DALL2542412R0302
- DALL2542411R0302
- Ambidextrous / hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Cuff style:
- Short
- Short
- Short
- Short
- Cuff length:
- 100 mm
- 100 mm
- 100 mm
- 100 mm
- Food contact approved:
- +
- +
- +
- +