Order Entry
China
ContactUsLinkComponent
Potassium hydroxide 45% in aqueous solution, AR®, Macron Fine Chemicals™
Corrosive
Irritant
Potassium hydroxide 45% in aqueous solution, AR®, Macron Fine Chemicals™
Catalog # MACR6671-04
CAS Number:  
Corrosive
Irritant
Potassium hydroxide 45% in aqueous solution, AR®, Macron Fine Chemicals™
Catalog # MACR6671-04
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Plastic receptacle
  • Pk:
    4 L
  • Seq:
    5162

Specifications

Specification Test Results

  • For Laboratory,Research,or Manufacturing Use:
    ≥45%
  • Assay (KOH):
    ≥45 %
  • Chloride (Cl):
    ≤0.002 %
  • Sulfate (SO₄):
    ≤0.001 %
  • Phosphate (PO₄):
    ≤0.0002 %
  • Ammonium Hydroxide Precipitate:
    ≤0.005 %
  • Iron (Fe):
    ≤0.0002 %
  • Heavy Metals (as Ag):
    ≤0.0005 %
  • Nickel (Ni):
    ≤0.0005 %
  • Sodium (Na):
    ≤0.02 %
  • Potassium Carbonate (K₂CO₃):
    ≤0.2 %
  • Appearance:
    Passes Test
  • Identification:
    Passes Test

Specification Test Results

About this item

 :
 :56.11 g/mol
Storage Temperature:Ambient
EINECS:215-181-3
UN:1814
ADR:8,II
 :GHS05
 :GHS07
CAS Number:1310-58-3
Formula:KOH
MDL Number:MFCD00003553