Order Entry
China
ContactUsLinkComponent
Diethyl ether ≥98% (by GC) stabilised, AR® ACS, Macron Fine Chemicals™
Flammable
Irritant
Health
Diethyl ether ≥98% (by GC) stabilised, AR® ACS, Macron Fine Chemicals™
Certificates
undefined
Flammable
Irritant
Health
Diethyl ether ≥98% (by GC) stabilised, AR® ACS, Macron Fine Chemicals™

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specification Test Results

  • ACS - Assay ((C₂H₅)₂O) (by GC, exclusive of preservative):
    ≥98.0 %
  • Appearance (clear, colorless liquid):
    Passes Test
  • ACS - Carbonyl Compounds (as HCHO) (by polarography):
    ≤0.001 %
  • ACS - Color (APHA):
    ≤10
  • ACS - Peroxide (as H₂O₂):
    ≤1 ppm
  • ACS - Preservative (BHT):
    ≥0.00005 %
  • ACS - Residue after Evaporation:
    ≤0.001 %
  • ACS - Titrable Acid (µeq/g):
    ≤0.2

Specification Test Results