1,1,1,3,3,3-Hexafluoro-2-propanol ≥99.8% eluent additive, for LC-MS, Fluka™
HONC42060-10X1ML
:
- Pack type:Ampoule
- Pk:10 mL
- Seq:2076
- Environmentally Preferable:
- Pk Info:10 x 1 ml
- Assay (GC):min. 99.8 %
- Suitability for the LC-MS:complying
- Non-volatile matter:Max. 5 ppm
- Water (Karl Fischer):Max. 200 ppm
- Absorbance at 240 nm:Max. 0.01
- Absorbance at 200 nm:Max. 0.3
- Aluminium (Al):Max. 0.5 ppm
- Barium (Ba):Max. 0.1 ppm
- Bismuth (Bi):Max. 0.1 ppm
- Calcium (Ca):Max. 0.5 ppm
- Cadmium (Cd):Max. 0.05 ppm
- Cobalt (Co):Max. 0.02 ppm
- Chromium (Cr):Max. 0.02 ppm
- Copper (Cu):Max. 0.02 ppm
- Iron (Fe):Max. 0.1 ppm
- Potassium (K):Max. 0.5 ppm
- Lithium (Li):Max. 0.1 ppm
- Magnesium (Mg):Max. 0.1 ppm
- Manganese (Mn):Max. 0.02 ppm
- Molybdenum (Mo):Max. 0.1 ppm
- Sodium (Na):Max. 1 ppm
- Nickel (Ni):Max. 0.02 ppm
- Lead (Pb):Max. 0.1 ppm
- Strontium (Sr):Max. 0.1 ppm
- Zinc (Zn):Max. 0.1 ppm
: 1,1,1,3,3,3-Hexafluoroisopropyl alcohol, Hexafluoro-2-propanol, Hexafluoroisopropanol, 1,1,1,3,3,3-Hexafluoropropan-2-ol, HFIP
:
:59 °C
:-4 °C
:–4 °C
:1.596 g/cm³ (25 °C)
:>100 °C
:Ambient
:213-059-4
:3265
:8,II
:168.04 g/mol
:GHS07
:GHS05
:920-66-1
:C₃H₂F₆O
:MFCD00011651