Order Entry
China
ContactUsLinkComponent
ALCH, CHROMASOLV™ for HPLC (denatured with 5% isopropanol and 5% methanol), Riedel-de Haën™
Irritant
Health
Flammable
ALCH, CHROMASOLV™ for HPLC (denatured with 5% isopropanol and 5% methanol), Riedel-de Haën™
Catalog # HONC270741-4X4L
Supplier:  HONEYWELL
CAS Number:  
Irritant
Health
Flammable
ALCH, CHROMASOLV™ for HPLC (denatured with 5% isopropanol and 5% methanol), Riedel-de Haën™
Catalog # HONC270741-4X4L
Supplier:  HONEYWELL
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    16 L
  • Seq:
    5254

Specifications

Specification Test Results

  • Identity (IR):
    complying
  • Assay (GC):
    Min. 89.0 %
  • Non-volatile matter:
    Max. 0.0005 %
  • Water (Karl Fischer):
    Max. 0.1 %
  • Methanol (GC):
    3.0 - 5.0 %
  • 2-propanol (GC):
    4.0 - 6.0 %
  • Absorbance at 400 nm:
    Max. 0.01
  • Absorbance at 300 nm:
    Max. 0.01
  • Absorbance at 255 nm:
    Max. 0.04
  • Absorbance at 220 nm:
    Max. 0.35
  • Absorbance at 210 nm:
    Max. 0.65
  • Absorbance at 205 nm:
    Max. 1.00

Specification Test Results

About this item

 :
Boiling Pt:78.32 °C (main component)
Melting Pt:–114.1 °C (main component)
Density:0.7890 g/cm³ (20 °C)
Flash Pt:15 °C (main component)
Storage Temperature:Ambient
UN:1987
ADR:3,II
 :GHS07
 :GHS08
 :GHS02
CAS Number:64-17-5
MDL Number:MFCD00003568