Size exclusion chromatography columns, InfinityLab AdvanceBio
Supplier: AGILENT HEWLETT PACKARD
Certificates
About this item
These columns are suitable for aggregation analysis via size exclusion chromatography.
- Increased analytical speed to help you meet vital deadlines
- Increased resolution for more accurate quantitation
- Increased sensitivity to quantitate aggregates, even at low levels
- Increased reproducibility to eliminate rework
AdvanceBio SEC columns deliver accurate, precise quantitation for mAb aggregation and protein SEC analysis.
In addition, AdvanceBio SEC columns improve lab productivity by providing robust, reliable methods that eliminate sample re-analyses. You can also easily transfer methods to other locations, including QA/QC, reducing the risk of late-stage candidate and batch failures.
Specifications
- Catalog No:
- HEWLPL1180-5301
- HEWLPL1180-1301
- Description:
- 300 Å
- 300 Å
- Max. pressure:
- 400 bar
- 400 bar
- pH range:
- 2 – 8,5
- 2 – 8,5