跳转到 页眉
跳转到 主要内容
跳转到 页脚
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
¥0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
China
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
Pharmaceutical secondary standard, lycopene, (CRM), Supelco®
Pharmaceutical secondary standard, lycopene, (CRM), Supelco®
Pharmaceutical secondary standard, lycopene, (CRM), Supelco®
Catalog # SUPLPHR1770-500MG
Supplier:
SUPELCO
CAS Number:
展开图片
Pharmaceutical secondary standard, lycopene, (CRM), Supelco®
Catalog # SUPLPHR1770-500MG
Supplier:
SUPELCO
CAS Number:
Specifications
Pk:
500 mg
CAS No.:
502-65-8
Description:
Lycopene
Description:
Lycopene
Specifications
About this item
Product Details
Product Details