Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
¥0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
China
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
Pharmaceutical secondary standard, Choline Bitartrate, (CRM), Supelco®
Pharmaceutical secondary standard, Choline Bitartrate, (CRM), Supelco®
Pharmaceutical secondary standard, Choline Bitartrate, (CRM), Supelco®
Catalog # SUPLPHR1460-500MG
Supplier:
SUPELCO
CAS Number:
Expand image
Pharmaceutical secondary standard, Choline Bitartrate, (CRM), Supelco®
Catalog # SUPLPHR1460-500MG
Supplier:
SUPELCO
CAS Number:
Specifications
Pk:
500 mg
CAS No.:
87-67-2
Description:
Choline Bitartrate
Description:
Choline Bitartrate
Specifications
About this item
Product Details
Product Details