Supelco® Drug and metabolite standards, Testosterone-2, 2,3,4-13C3 solution, (CRM)
目录编号 SUPLT-070-1ML
Supplier: SUPELCO
CAS Number:
Specifications
- Concentration:100 µg/ml in acetonitrile
- Pk:1 mL
- Standards:Certified reference material
- Description:Testosterone-2, 2,3,4-13C3 solution
- Pk Info:ampoule of 1 ml