Drug and metabolite standards, Caffeine solution, (CRM), Supelco®
Catalog # SUPLC-051-1ML
Supplier: SUPELCO
CAS Number:
Specifications
- Concentration:1,0 mg/ml in methanol
- Pk:1 mL
- Standards:Certified reference material
- CAS No.:58-08-2
- Description:Caffeine solution
- Description:Caffeine solution
- Pk Info:ampoule of 1 ml