Specifications
- Weight:0,226 kg
- Temperature range:–50,000...150,000 °C
- Description:Digital thermometer with Probe
- W×D×H:88,9×139,9×32.0 mm
- Accuracy:±0,05 °C between 0,0...100 °C
- Pk:1
Specifications
About this item
Ideal for measuring temperatures in cuvettes, gas systems, reagents, and chemical solutions, or for routine measurements.
- Units read °C or °F at the touch of a button
- Compact and portable
- Resolution of 0,001°
- High-contrast LCD features 12,7 mm
The Data Acquisition System captures, displays, and stores readings in Windows®. Data can be read as-is or imported into databases, spreadsheets, word processors, and statistical programs.
Readings are updated three times a second. Features include hold button, minimum/maximum memories, and 50 data logger memories. Units operate on 9 V alkaline battery (included) or optional 115 V adapter/recharger.Thermometer comes with a standard quick-response, stainless steel triple purpose probe that features a 1,5 m (5') cable and can be used with liquids, semi-solids or air/gas.
Ideal for foods, pharmaceuticals, petroleum products, gas systems, soil, ovens, and more.