VWR® DuPont™ Tyvek® IsoClean®, Frocks
Supplier: VWR BRAND PRIVATE LABEL USA
Certificates
About this item
Frocks provide a high level of protection in cleanroom environments.
- Chemically and biologically inert
- Low linting
- Static-free until laundered
These limited-use frocks with five-snap front feature a bound neck and bound seams. All garments are pocketless.
Specifications
- Catalog No:
- VWRU80200-610
- VWRU80200-608
- Closure type:
- Buttons
- Buttons
- Pockets:
- Without pockets
- Without pockets
- Garment application:
- Laboratory
- Laboratory