Formalin 10%, buffered
Catalog # VWRU89370-094
Supplier: VWR BRAND PRIVATE LABEL USA
CAS Number:
Specifications
- Pk:1
- Seq:0001
- Environmentally Preferable:
Specifications
About this item
Synonyms: Paraformaldehyde aqueous solution, Formaldehyde, Methylaldehyde, Formol
:
:30.03 g/mol
Storage Temperature:Ambient
CAS Number:50-00-0
Formula:CH₂O
MDL Number:MFCD00003274