Order Entry
China
ContactUsLinkComponent
VWR®Traceable®, Programmable time switch, outlet controller
VWR®Traceable®, Programmable time switch, outlet controller
Catalog # VWRU61161-343
VWR®Traceable®, Programmable time switch, outlet controller
Catalog # VWRU61161-343

Specifications

  • Description:
    Programmable time switch with digital outlet controller
  • W×D×H:
    102×89×51 mm
  • Accuracy:
    0,01 %
  • Pk:
    1

Specifications

About this item

This versatile and economical controller programs equipment on and off at any minute of any day. A fail-safe memory permits programming 14 different switchings per day, up to 196 switchings per week.

  • Accuracy: 0,001%
  • Timing capacity: 1 min to 168 hours
  • Switching capacity: 125 V, 50/60 Hz, 1850 W, 15 A
  • Horsepower: 1/3
  • Manual override, backup battery, random interval setting

The AM/PM day programs allow repeating same cycle everyday, weekdays only, weekends only or any selected days.

Certifications: CSA and NTRL-C-rated.
Ordering information: Supplied with two batteries and Traceable® certificate.