VWR®, Channel connector
Catalog # VWRU60079-012
Supplier: VWR BRAND PRIVATE LABEL USA
Specifications
- Description:Channel connector
- Pk:1
Specifications
About this item
Special connector fastens lab frame rod to 41 mm steel channels.
- Easy to adjust
- Securely fastens lab frame rod to steen channels
- Available individually or in bulk
Loosen the connector or slide up or down for easy adjustment of rod position.