Polypropylene beakers with a pouring spout and graduations printed directly on them.
- Stackable
- Working temperatures of 0 to 121 °C
Colour: Natural
: Do not stack products while autoclaving.
Polypropylene beakers with a pouring spout and graduations printed directly on them.
Colour: Natural