Order Entry
China
ContactUsLinkComponent
VWR® Volumetric Flask, Clear Glass, Class A, Serialised, with Penny Head Glass Stopper
VWR® Volumetric Flask, Clear Glass, Class A, Serialised, with Penny Head Glass Stopper
Catalog # VWRU10124-376
undefined
VWR® Volumetric Flask, Clear Glass, Class A, Serialised, with Penny Head Glass Stopper
Catalog # VWRU10124-376
Specifications
  • Capacity:
    1000 ml
  • Description:
    Volumetric flask
  • Pk:
    1
  • Tolerance:
    0,3 ml
Specifications
About this item
Made from borosilicate 3.3 expansion glass.
Flasks are serialised and supplied with glass penny head stopper.