VWR®, Digital Timer, One Channel, with Memory and Clock
VWRU82023-858
- Accuracy:3 s
- Pk:1
- W×H:70×51 mm
- Timing capacity:23 h, 59 min, 59 s
Mounting options include lay flat on a bench, use the flip-open stand for a vertical view, carry it on a lab coat with the convenient pocket clip, or use the magnetic back for metal surface attachment.
- Clock feature
- Last time set memory
- Multiple mounting options
Features a Certificate of Calibration and individual serial number, these electronic timers are tested and calibrated in H-B’s exclusive triple accredited/registered ISO/IEC 17025:2005, A2LA accredited laboratory (Cert #2448.01), against equipment whose calibration is traceable to NIST.
: RoHS compliant.
: Requires one LR44 battery (included).