Order Entry
China
ContactUsLinkComponent
VWR®, Filtering Flasks, Graduated, Heavy Wall, with Glass Connector
  VWRU10545-860
undefined
VWR®, Filtering Flasks, Graduated, Heavy Wall, with Glass Connector
  VWRU10545-860
  • Capacity:
    2000 ml
  • Seq:
    07
  • Pk:
    1
These flasks with glass connector feature a heavy wall construction which can withstand high vacuum.
  • White enamel markings
  • Chemical resistant
Manufactured from ASTM E438 Type 1 Class A borosilicate 33 expansion glass.
 : Tubing not included.