Strips of 8 PCR tubes with attached flat individual caps
: BRAND
PP. Eight connected 0,2 or 0,15 ml PCR tubes with attached flat individual caps. The tubes are held together by three connectors for increased rigidity.
- Cap offers protection against contamination
- Highly transparent cap makes them ideal for real-time PCR (qPCR)
- Available in standard or low profile versions
- Free from DNase, RNase and DNA
Colour: Clear or white
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.