0
¥0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
China
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Spatulas
Pharmacist's spatulas
Pharmacist's spatulas
Pharmacist's spatulas
Supplier:
BOCHEM
Certificates
Expand image
Pharmacist's spatulas
Supplier:
BOCHEM
About this item
These spatulas feature a stainless steel blade and a PVC handle.
Suitable for dishwasher
Autoclavable