Order Entry
China
ContactUsLinkComponent
N,N-Dimethylformamide ≥99.9%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Flammable
Health
Irritant
N,N-Dimethylformamide ≥99.9%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Catalog # HONC270547-4X4L
Supplier:  HONEYWELL
CAS Number:  
undefined
Flammable
Health
Irritant
N,N-Dimethylformamide ≥99.9%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Catalog # HONC270547-4X4L
Supplier:  HONEYWELL
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    16 L
  • Seq:
    5254

Specifications

Specification Test Results

  • IR spectrum:
    complying
  • Assay (GC):
    Min. 99.90 %
  • Non-volatile matter:
    Max. 0.0005 %
  • Water (Karl Fischer):
    Max. 0.03 %
  • Absorbance at 270 nm:
    Max. 1.0
  • Absorbance at 275 nm:
    Max. 0.30
  • Absorbance at 295 nm:
    Max. 0.10
  • Absorbance at 310 nm:
    Max. 0.05
  • Absorbance at 340 nm:
    Max. 0.01
  • Absorbance at 400 nm:
    Max. 0.01
  • Explosion Limit:
    15.2 %
  • Vapor Pressure:
    2.7 mmHg ( 20 °C)
  • Refractive Index:
    n20/D 1.430 (lit.)

Specification Test Results

About this item

 :
 :73.09 g/mol
Merck Index:13,03269
Boiling Pt:152-153 °C (1.013 hPa)
Boiling Pt:152…153 °C (1.013 hPa)
Melting Pt:-61 °C
Melting Pt:–61 °C
Density:0.94 g/cm³ (20 °C)
Flash Pt:57.5 °C (1.013.25 hPa)
Storage Temperature:Ambient
EINECS:200-679-5
UN:2265
ADR:3,III
REACH:01-2119475605-32
 :GHS02
 :GHS08
 :GHS07
CAS Number:68-12-2
Formula:HCON(CH₃)₂
MDL Number:MFCD00003284