


Tetrahydrofuran ≥99.9% inhibitor-free, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Catalog # HONC34865-1L
Supplier: HONEYWELL
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Pack type:Glass bottle
- Pk:1 L
- Seq:5037
Specifications
Specification Test Results
- Assay (GC):Min. 99.90 %
- IR spectrum:complying
- Non-volatile matter:Max. 0.0005 %
- Water (Karl Fischer):Max. 0.05 %
- Free acid (as CH₃COOH):Max. 0.002 %
- Peroxides (as H₂O₂):Max. 0.05 %
- APHA:Max. 10
- Absorbance at 400 nm:Max. 0.005
- Absorbance at 350 nm:Max. 0.005
- Absorbance at 315 nm:Max. 0.0044
- Absorbance at 300 nm:Max. 0.02
- Absorbance at 275 nm:Max. 0.046
- Absorbance at 250 nm:Max. 0.18
- Absorbance at 245 nm:Max. 0.26
- Absorbance at 212 nm:Max. 1.0
- Explosion Limit:11.8 %
- Vapor Density:2.5 (vs air)
- Refractive Index:n20/D 1.407 (lit.)
- Autoignition Temperature:610 °F
Specification Test Results
About this item
:
:72.11 g/mol
Merck Index:12,09351
Boiling Pt:65 °C (1.013 hPa)
Melting Pt:–108 °C
Density:0.89 g/cm³ (20 °C)
Flash Pt:–21 °C
Storage Temperature:Ambient
EINECS:203-726-8
UN:2056
ADR:3,II
REACH:05-2114615188-48
:GHS07
:GHS08
:GHS02
CAS Number:109-99-9
Formula:C₄H₈O
MDL Number:MFCD00005356