
Lithium carbonate ≥99.0% (by titrimetric analysis), Puriss. p.a. ACS for microscopy, Fluka™
Catalog # HONC62470-100G
Supplier: HONEYWELL
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:100 g
- Seq:3401
Specifications
Specification Test Results
- Assay:99.0 - 101.0 %
- Insoluble in HCl:Max. 0.01 %
- Calcium (Ca):Max. 50 ppm
- Cadmium (Cd):Max. 5 ppm
- Cobalt (Co):Max. 5 ppm
- Chromium (Cr):Max. 5 ppm
- Copper (Cu):Max. 5 ppm
- Iron (Fe):Max. 5 ppm
- Potassium (K):Max. 50 ppm
- Magnesium (Mg):Max. 10 ppm
- Manganese (Mn):Max. 5 ppm
- Sodium (Na):Max. 50 ppm
- Nickel (Ni):Max. 5 ppm
- Lead (Pb):Max. 5 ppm
- Zinc (Zn):Max. 5 ppm
- Heavy metals (ICP) ACS:Max. 0.002 %
- S-compounds (as SO₄):Max. 50 ppm
- Chloride (Cl):Max. 20 ppm
- Ammonium (NH₄):Max. 5 ppm
- Nitrate (NO₃):Max. 5 ppm
Specification Test Results
About this item
:
:73.89 g/mol
Merck Index:13,05549
Boiling Pt:1.300 °C
Melting Pt:720 °C
Density:2.110 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:209-062-5
:GHS07
CAS Number:554-13-2
Formula:Li₂CO₃
MDL Number:MFCD00011084