Methanol ≥99.9%, CHROMASOLV™ for HPLC, Riedel-de Haën™
Supplier: HONEYWELL
Certificates
About this item
Synonyms: Wood alcohol, Methyl alcohol, Hydroxy methane, Carbinol
CAS Number:67-56-1
:
Formula:CH₃OH
MDL Number:MFCD00004595
:32.04 g/mol
Merck Index:13,05984
Boiling Pt:64…65 °C (1.013 hPa)
Melting Pt:–98 °C
Density:0.79 g/cm³ (20 °C)
Flash Pt:11 °C
Storage Temperature:Ambient
EINECS:200-659-6
UN:1230
ADR:3,II
REACH:01-2119433307-44
:GHS08
:GHS02
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Assay (GC):Min. 99.90 %
- Non-volatile matter:Max. 0.0005 %
- Water (Karl Fischer):Max. 0.03 %
- Absorbance at 400 nm:Max. 0.01
- Absorbance 280 nm:Max. 0.01
- Absorbance at 260 nm:Max. 0.04
- Absobance at 254 nm:Max. 0.02
- Absorbance at 240 nm:Max. 0.07
- Absorbance at 235 nm:Max. 0.10
- Absorbance at 230 nm:Max. 0.15
- Absorbance at 220 nm:Max. 0.30
- Absorbance at 210 nm:Max. 0.60
- Absorbance at 205 nm:Max. 1.00
- Absorbance (JP)(Val.Ref):complying
- Fluorescence (chinin) at 254 nm:Max. 1 ppb
- Fluorescence (chinin) at 365 nm:Max. 1 ppb
- APHA:Max. 10
- Vapor Pressure:97.68 mmHg ( 20 °C)
- Vapor Density:1.11 (vs air)