Order Entry
China
ContactUsLinkComponent
Acetonitrile ≥99.9%, CHROMASOLV™, gradient grade for HPLC, Riedel-de Haën™
Health
Irritant
Flammable
Acetonitrile ≥99.9%, CHROMASOLV™, gradient grade for HPLC, Riedel-de Haën™
Supplier:  HONEYWELL
   Sustainable Options Available
Certificates
undefined
Health
Irritant
Flammable
Acetonitrile ≥99.9%, CHROMASOLV™, gradient grade for HPLC, Riedel-de Haën™
Supplier:  HONEYWELL
   Sustainable Options Available

Specifications

  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended by default

Specifications

Specification Test Results

  • Assay (GC):
    Min. 99.90 %
  • Refractive index (n 20/D):
    1.3435 - 1.3445
  • Non-volatile matter:
    Max. 0.0005 %
  • Water (Karl Fischer):
    Max. 0.02 %
  • Free acid (as CH₃COOH):
    Max. 0.001 %
  • Free alkali (as NH₃):
    Max. 0.0002 %
  • Absorbance at 400 nm:
    Max. 0.0044
  • Absorbance at 250 nm:
    Max. 0.0044
  • Absorbance at 235 nm:
    Max. 0.0044
  • Absorbance at 230 nm:
    Max. 0.0044
  • Absorbance at 200 nm:
    Max. 0.032
  • Absorbance at 195 nm:
    Max. 0.12
  • Fluorescence (chinin) at 254 nm:
    Max. 1 ppb
  • Fluorescence (chinin) at 365 nm:
    Max. 0.5 ppb
  • HPLC Gradient at 210 nm:
    Max. 3 mAU
  • HPLC Gradient at 254 nm:
    Max. 0.5 mAU
  • Baseline drift at 210 nm:
    Max. 15 mAU
  • Refractive Index:
    n20/D 1.344 (lit.)
  • Measuring Range:
    ≤15 mAU baseline drift, (210 nm)
  • Autoignition Temperature:
    973 °F

Specification Test Results

Related Information