Barium sulphate 97.5-100.5% USP suitable for use as excipient, Macron Fine Chemicals™
Catalog # MACR4518-26
Supplier: MACRON AVANTOR BRAND
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Pack type:Fibre drum
- Pk:250 lb
- Seq:7215
Specifications
Specification Test Results
- Bulk Pharmaceutical Chemical:97.5 - 100.5%
- USP - Assay (BaSO₄):97.5 - 100.5 %
- USP - Acid-Soluble Substances:≤0.3 %
- USP - Bulk (fl. oz/lb):20 - 31
- USP - Identification A:Passes Test
- USP - Identification B:Passes Test
- USP - pH (10% Suspension):3.5 - 10.0
- USP - Soluble Barium Salts:≤0.001 %
- USP - Sulfide (S):≤0.5 ppm
- Suspension Test for Dirt:Passes Test
Specification Test Results
About this item
:
:233.39 g/mol
Merck Index:13,00997
Boiling Pt:1600 °C (1013 hPa)
Melting Pt:1345 °C
Density:4.5 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:231-784-4
CAS Number:7727-43-7
Formula:BaSO₄
MDL Number:MFCD00003455