订单输入
China
ContactUsLinkComponent
甲苯 ≥99.7% (通过GC), ChromAR® 用于液相色谱, 用于紫外分光光度法, Macron Fine Chemicals™
 
 
 
甲苯 ≥99.7% (通过GC), ChromAR® 用于液相色谱, 用于紫外分光光度法, Macron Fine Chemicals™
  MACR4483-10
 :  
undefined
 
 
 
甲苯 ≥99.7% (通过GC), ChromAR® 用于液相色谱, 用于紫外分光光度法, Macron Fine Chemicals™
  MACR4483-10
 :  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

 

  • 包装类型:
    Glass bottle, amber, narrow mouth
  • 包装规格:
    4 L
  • 顺序:
    5162

 

 

  • For Laboratory,Research,or Manufacturing Use:
  • Assay (C₆H₅CH₃) (by GC):
    ≥99.7 %
  • Color (APHA):
    ≤10
  • Residue after Evaporation:
    ≤4 ppm
  • Substances Darkened by H₂SO₄:
    Passes Test
  • Sulfur Compounds (as S):
    ≤0.003 %
  • Water (H₂O)(by KF, coulometric):
    ≤0.03 %
  • Optical Absorbance (1-cm path vs water)::
    Passes Test
  • Absorbance - 285 nm:
    ≤1.000
  • Absorbance - 286 nm:
    ≤0.700
  • Absorbance - 288 nm:
    ≤0.400
  • Absorbance - 293 nm:
    ≤0.200
  • Absorbance - 300 nm:
    ≤0.104
  • Absorbance - 310 nm:
    ≤0.050
  • Absorbance - 335 nm:
    ≤0.020
  • Absorbance 350 - 400 nm:
    ≤0.010

 

 

 :
 :92.14 g/mol
 :13,09607
 :110,6 °C (1013 hPa)
 :–95 °C
 :0,867 g/cm³ (20 °C)
 :4 °C
 :Ambient
 :203-625-9
 :1294
 :3,II
 :05-2114615130-69
 :GHS02
 :GHS08
 :GHS07
 :108-88-3
 :C₆H₅CH₃
 :MFCD00008512