DL-二硫苏糖醇 ≥99.0%, Biotech Reagent, J.T.Baker®
目录编号 BAKRF781-05
供应商: Avantor
CAS 编号:
规格
- 包装类型:Poly bottle, white, blue T/E cap
- 包装规格:1 kg
- 顺序:4024
- Environmentally Preferable:
规格
规格测试结果
- Keep refrigerated between 2-8°C (36-46°F) and store under nitrogen.:≥99.0%
- CAUTION: For Manufacturing, processing or repackaging:≤0.060
- Assay (HSCH₂(CHOH)₂CH₂SH):≥99.0 %
- Appearance:Passes Test
- Absorbance of a 0.1M Solution (1-cm path vs water) at - 280 nm:≤0.060
- Absorbance of a 0.1M Solution (1-cm path vs water) at - 260 nm:≤0.400
- Infrared Spectrum:Passes Test
规格测试结果
关于此项目
同义词: 1,4-二巯基苏糖醇, 克莱兰氏试剂, DTT
:
:154.25 g/mol
沸点:∼125 °C (1013 hPa)
熔点:42…44 °C
闪点:>110 °C
储存温度:Refrigerator
EINECS:222-468-7
:GHS07
CAS 编号:3483-12-3
:27565-41-9
化学式:C₄H₁₀O₂S₂
MDL 编号:MFCD00004877