Order Entry
China
ContactUsLinkComponent
Equivalence, Column Coupler
Equivalence, Column Coupler
Catalog # ACETEQV-C0001
Supplier:  Avantor
Equivalence, Column Coupler
Catalog # ACETEQV-C0001
Supplier:  Avantor
Specifications
  • Description:
    Column coupler
  • Pk:
    1
Specifications
About this item
This column coupler is mainly used with guard column cartridges for narrow bore columns 1,0 mm ID and semi-prep cartridge holder.