Tris Buffered Saline, 10X concentrate solution, Sigma-Aldrich®
Catalog # SIALT5912-1L
Supplier: SIGMAALDRICH
CAS Number:
Specifications
- Pk:1 L
- Description:TRIS buffered saline, 10X, pH 7,4
Specifications
About this item
Washing buffer for alkaline phosphatase or peroxidase conjugates in Western blotting.
- 0,2 μm filtered
- Store at 2 to 8 °C
Working solution will contain 20 mM Tris and 0,9% NaCl. Tris Buffered Saline has been used as washing buffer for electrodes. It can be used to wash microtiter plates for western blot experiments and screening methods.