Order Entry
China
ContactUsLinkComponent
Lithiumbis(trimethylsilyl)amide 95%
 
 
Lithiumbis(trimethylsilyl)amide 95%
  ACRO338140100
 :  
undefined
 
 
Lithiumbis(trimethylsilyl)amide 95%
  ACRO338140100
 :  

 

  • Pack type:
    Glass bottle for solids
  • Pk:
    10 g
  • Seq:
    3236

 

 

  • Appearance:
    Yellow Crystalline powder and/or chunks
  • Infrared spectrum:
    Conforms
  • Titration with HClO4:
    ≥94 %

 

 

 :
 :167.33 g/mol
 :114…116 °C
 :105 - 107 °C
 :0.891 g/cm³
 :Ambient
 :223-725-6
 :2925
 :4.1,II
 :GHS02
 :GHS05
 :4039-32-1
 :C₆H₁₈LiNSi₂
 :MFCD00008261