L(+)-Ascorbic acid 99.0-100.5%, fine-granular USP, FCC 20 - 80 Mesh, J.T.Baker®
Supplier: Avantor
Certificates
About this item
CAS Number:50-81-7
:
Formula:C₆H₈O₆
MDL Number:MFCD00064328
:176.13 g/mol
Merck Index:13,00837
Boiling Pt:553 °C (1013 hPa)
Melting Pt:190…192 °C
Density:1.65 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-066-2
REACH:exempt
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specification Test Results
- Meets F.C.C. Requirements:99.0 - 100.5%
- GMP Manufactured Product:Passes Test
- Meets U.S.P Requirements:Passes Test
- Food GMP Manufactured Product:20.5 - 21.5°
- CAUTION: For Manufacturing, processing or repackaging:≤0.1%
- Bulk Pharmaceutical Chemical:99.0 - 100.5%
- Bulk Food Chemical:Passes Test
- USP - Assay (C₆H₈O₆):99.0 - 100.5 %
- USP - Identification A:Passes Test
- USP - Identification B:Passes Test
- USP - Specific Rotation [Å]²⁵ ^D (+):20.5 - 21.5 Degree
- USP - Residue on Ignition:≤0.1 %
- FCC - Assay (C₆H₈O₆):99.0 - 100.5 %
- FCC - Identification A:Passes Test
- FCC - Identification B:Passes Test
- FCC - Lead (Pb):≤2 mg/kg
- FCC - Specific Rotation [Å]²⁵ ^D (+):20.5 - 21.5 Degree
- FCC - Residue on Ignition:≤0.1 %
- Arsenic (As):≤3 ppm
- Mesh - Thru U.S. No. 20 Sieve:≥100 %
- Mesh - Thru U.S. No. 30 Sieve:≥95 %
- Mesh - Thru U.S. No. 80 Sieve:≤20 %