Order Entry
China
ContactUsLinkComponent
Phosphate buffered saline (PBS), Sigma-Aldrich®
Phosphate buffered saline (PBS), Sigma-Aldrich®
Catalog # SIALP3563-10PAK
Supplier:  SIGMAALDRICH
CAS Number:  
Phosphate buffered saline (PBS), Sigma-Aldrich®
Catalog # SIALP3563-10PAK
Supplier:  SIGMAALDRICH
CAS Number:  
Specifications
  • Pack type:
    Foil pouches
  • Pk:
    1
  • Description:
    Phosphate buffered saline
  • Seq:
    01
  • M-Clarity quality level:
    MQ200
  • Pk Info:
    1× 10 pouches
Specifications
About this item
Contents of one pouch, when dissolved in one liter of distilled or deionised water, will yield 0,01 M phosphate buffered saline (NaCl 0,138 M; KCl - 0,0027 M); TWEEN® 20 - 0,05%, pH 7,4, at 25 °C.