Order Entry
China
ContactUsLinkComponent
Manganese(II) chloride, anhydrous 99+% 80 mesh
Corrosive
Irritant
Health
Environment
Manganese(II) chloride, anhydrous 99+% 80 mesh
Certificates
undefined
Corrosive
Irritant
Health
Environment
Manganese(II) chloride, anhydrous 99+% 80 mesh

Specification Test Results

  • Appearance:
    Pink Powder
  • Titration Complexometric:
    ≥99.0 %
  • Solubility:
    (10 % in dilute HCl) Clear solution
  • Particle size:
    -80 mesh

Specification Test Results

Related Information