Order Entry
China
ContactUsLinkComponent
 
Tungsten(VI) chloride 99% 20 mesh
 
undefined
Tungsten(VI) chloride 99% 20 mesh

 

  • Environmentally Preferable:

 

 

  • Appearance:
    Grey to dark blue to violet to black Powder
  • Purity:
    ≥98.5 %