Titanium(IV) ethanolate (33 - 35% TiO₂)
: Titanium (IV) ethoxide, Titanium tetraethanolate, Tetraethyl orthotitanate
:3087-36-3
:
:C₈H₂₀O₄Ti
:MFCD00009071
:150…152 °C (10 mmHg)
:–40 °C
:1.09 g/cm³
:228.12 g/mol
:33 °C
:Ambient
:221-410-8
:1993
:3 III
:GHS02
:GHS07
- Environmentally Preferable:
- Appearance:Clear colorless to yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5030 to 1.5070 (20°C, 589 nm)
- Titration Complexometric:33 to 35 % (TiO2)