Order Entry
China
ContactUsLinkComponent
 
 
Diyttrium trioxide 99.99% (trace metals basis)
 
undefined
 
Diyttrium trioxide 99.99% (trace metals basis)

 

  • Appearance:
    White Powder
  • Assay:
    ≥99.99 % (Y2O3) TREO base
  • Particle size:
    2 to 10 µm (d50)
  • Total conc. of imp.as REO:
    ≤100 ppm
  • Loss on ignition:
    ≤1.0 % (1000°C) Material base