Order Entry
China
ContactUsLinkComponent
 
 
Diyttrium trioxide 99.999% (trace metals basis)
 
undefined
 
Diyttrium trioxide 99.999% (trace metals basis)

 

  • Appearance:
    White Powder
  • Assay:
    99.999 % (Y2O3) On TREO base