Order Entry
China
ContactUsLinkComponent
Mercury(II) sulphate ACS
Mercury(II) sulphate ACS
Certificates
undefined
Mercury(II) sulphate ACS
Certificates

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • Appearance:
    White to off-white Crystalline powder
  • Titration with NH4SCN:
    ≥98.0 %
  • Chloride (Cl):
    ≤30 ppm
  • Nitrate (NO3):
    Passes test (limit about 0.003%)
  • Iron (Fe):
    ≤50 ppm
  • Mercurous Mercury (as Hg):
    ≤0.15 %
  • Residue after reduction:
    ≤0.02 %

Specification Test Results

Related Information