Order Entry
China
ContactUsLinkComponent
 
 
 
 
Copper(II) sulphate 98.5-100.5%, GenAR® USP for biotechnology, Macron Fine Chemicals™
  MACR7790-88
 :  
undefined
 
 
 
Copper(II) sulphate 98.5-100.5%, GenAR® USP for biotechnology, Macron Fine Chemicals™
  MACR7790-88
 :  

 

  • Pack type:
    PE pail, screw top
  • Pk:
    12 kg
  • Seq:
    4187

 

 

  • Preserve in tight containers. Store at 25°C, excursions permitted 15-30°C.:
    98.5 - 100.5 %
  • CAUTION: For Manufacturing, processing or repackaging:
    = 0.005 %
  • Bulk Pharmaceutical Chemical:
    Passes Test
  • USP - Assay (as CuSO₄):
    98.5 - 100.5 %
  • USP - Calcium (Ca):
    ≤0.005 %
  • Solubility (5 in 50):
    Passes Test
  • USP - Identification A:
    Passes Test
  • USP - Identification B:
    Passes Test
  • USP - Iron (Fe):
    ≤0.003 %
  • USP - Loss on Drying:
    33.0 - 36.5 %
  • USP - Nickel (Ni):
    ≤0.005 %
  • USP - Potassium (K):
    ≤0.01 %
  • USP - Sodium (Na):
    ≤0.02 %
  • Endotoxin Concentration (EU/g):
    ≤10
  • EP/BP - Assay:
    99.0 - 101.0 %
  • EP/BP - Identification A:
    Passes Test
  • EP/BP - Identification B:
    Passes Test
  • EP/BP - Identification C:
    Passes Test
  • EP/BP - Appearance of Solution:
    Passes Test
  • EP/BP - Chloride (Cl):
    ≤100 ppm
  • EP/BP - Iron (Fe):
    ≤100 ppm
  • EP/BP - Lead (Pb):
    ≤50 ppm
  • EP/BP - Loss on Drying:
    35.0 - 36.5 %

 

 

 :
 :159.61 g/mol
 :12,02722
 :200 °C
 :2.286 g/cm³ (15.6 °C)
 :Ambient
 :231-847-6
 :3288
 :6.1,III
 :Management of Change (MOC) category = RL
 :GHS05
 :GHS09
 :GHS08
 :7758-98-7
 :CuSO₄
 :MFCD00010981