Order Entry
China
ContactUsLinkComponent
Lutetium(III) oxide 99.99% (trace metals basis) -325 mesh
Irritant
Lutetium(III) oxide 99.99% (trace metals basis) -325 mesh
Catalog # ACRO193400010
CAS Number:  
undefined
Irritant
Lutetium(III) oxide 99.99% (trace metals basis) -325 mesh
Catalog # ACRO193400010
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    White Powder
  • Assay:
    ≥99.94 %

Specification Test Results

About this item

 :
 :397.93 g/mol
Melting Pt:2510 °C
Density:9.42 g/cm³
EINECS:234-764-3
 :GHS07
CAS Number:12032-20-1
Formula:Lu₂O₃
MDL Number:MFCD00011100