Supelco® Organic Reference Standard, Tetramethylsilane
SUPL87920-25ML
: SUPELCO
:
- Pk:25 mL
- Standards:Analytical standard, for NMR spectroscopy, ACS reagent
- CAS No.:75-76-3
- Description:Tetramethylsilane
- Storage conditions:2…8 °C
- Hazard class description:H224
- Shipping conditions:STD
- MDL No.:MFCD00008274
- M-Clarity quality level:MQ200
- Melting point:–99 °C (lit.)
- Formula:Si(CH₃)₄
- Molecular weight:88,22
- Density:0,648 g/ml at 25 °C (lit.)
- Flash point:–27 °C – Closed cup
- Boiling point:26 - 28 °C (lit.)
- GHS signal: GHS02
: P210