
(3-Glycidoxypropyl)trimethoxysilane for synthesis, Sigma-Aldrich®
Catalog # MERC8.41807.0100
Supplier: MERCK MILLIPORE
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:100 mL
- Seq:2440
Specifications
Specification Test Results
- Solubility:(20 °C) immiscible,(decomposition)
- Molar Mass:236.34 g/mol
- Boiling Point:290 °C (1013 hPa)
- Vapor Pressure:- 1.33 hPa (20 °C)
- Flash Point:122 °C
- Explosion Limit:0.43 %(V)
- Density:1.07 g/cm3 (20 °C)
- pH:6.8 (100 g/l, H2O)
- Ignition Point:400 °C
- Viscosity kinematic:3 mm2/s (25 °C)
Specification Test Results
About this item
:
:236.34 g/mol
Boiling Pt:299 °C (1013 hPa)
Melting Pt:0 °C
Density:1.07 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Flash Pt:122 °C
Storage Temperature:Ambient
EINECS:219-784-2
:GHS05
CAS Number:2530-83-8
Formula:C₉H₂₀O₅Si
MDL Number:MFCD00005144