Order Entry
China
ContactUsLinkComponent
Lead(II) oxide 99.9+% (trace metals basis), powder < 10 µm
Irritant
Health
Environment
Lead(II) oxide 99.9+% (trace metals basis), powder < 10 µm
Catalog # ACRO315855000
CAS Number:  
undefined
Irritant
Health
Environment
Lead(II) oxide 99.9+% (trace metals basis), powder < 10 µm
Catalog # ACRO315855000
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g
  • Seq:
    3645

Specifications

Specification Test Results

  • Appearance:
    Yellow to light orange Powder
  • Average particle size:
    ≤10 µm
  • Total trace metal impurities:
    ≤1000.0 ppm

Specification Test Results

About this item

 :
 :223.2 g/mol
Merck Index:13,05432
Boiling Pt:1470 °C
Melting Pt:886 °C
Density:4,53 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:233-245-9
UN:3077
ADR:9,III
 :GHS07
 :GHS08
 :GHS09
CAS Number:1317-36-8
Formula:PbO
MDL Number:MFCD00011164